FTIR Crystal Reference Guide
Issue
Determining the crystal materials best suited for infrared sampling needs
Environment
Fourier Transform Infrared systems
Resolution
Major considerations in choosing materials are:
- Frequency range,
- Solubility of the sample,
- Refractice Index,
- Reactivity with the sample, and
- Mechanical and thermal characteristics.
Use the attached Crystal Reference Guide to determine the crylstal materials best suited for infrared sampling needs.
Attachment(s)
| File | Last Modified |
|---|---|
| FTIR Crystal Reference Guide.pdf | December 14, 2021 |