How to Change the Battery on the Imaging ATR Accessory
Issue
The battery on the Imaging ATR Accessory pressure sensor needs replacement
Environment
Nicolet iN10
Nicolet Continuum
FT-IR Microscopes
Resolution
On the left side of the Pressure Sensor Housing, there is a battery holder that inserts into the unit. This is just in front of the silver knob.
- Pull on the end of the battery holder that sticks out slightly from the left side of the Pressure Sensor Housing and slide the holder from the unit.
- Remove the coin battery from the holder, observing the polarity of the battery.
- Insert the new coin battery into the holder in the proper orientation.
- Reinsert the holder into the Pressure Sensor Housing.
Analysis
Attachment(s)
| File | Last Modified |
|---|---|
| Pressure Sensor Holder with Battery holder.JPG | August 03, 2022 |
| Battery Holder.JPG | August 03, 2022 |