How to Disable the Windows 10 USB Selective Suspend Setting
Issue
Disabling the Windows 10 USB selective suspend setting
Environment
- FT-IR
- Windows 10
Resolution
Follow the steps in the attachment to disable the WIndows 10 USB selective suspend setting.
Analysis
Windows 10 includes the USB selective suspend feature that lets you automatically put USB devices in a very low-power state when they’re not actively in use. The feature was built in to prolong battery life on a laptop or tablet, but the feature may cause problems with communications with our FT-IR spectrometers connected to the PC.
Attachment(s)
| File | Last Modified |
|---|---|
| How to Disable the Windows 10 USB Selective Suspend Setting.pdf | December 15, 2021 |