Moving OMNIC data from old PC to new PC
Issue
Moving OMNIC data from old PC to new PC
Environment
- Omnic
Resolution
This explains a simple way to move your OMNIC data folders (Spectra, Macros, Quant, Libs, Param, Reports) from one computer to another.
Note: You must install OMNIC on the new PC first. This only moves your data, not the program.
-
On the old PC
-
Open File Explorer.
-
Go to: C:\My Documents\Omnic
-
Confirm you see folders like: Spectra, Macros, Quant, Libs, Param, Reports.
-
Right-click the Omnic folder and choose Copy.
-
Paste it to a USB drive, external drive, or network folder.
-
-
On the new PC
-
Install and run OMNIC once, then close it.
-
Open File Explorer and go to: C:\My Documents
-
Open the Omnic folder there, and copy the subfolders (Spectra, Macros, Quant, Libs, Param, Reports) into C:\My Documents\Omnic on the new PC.
-
When Windows asks about duplicates, choose to merge folders and do not overwrite files if you are unsure.
-
-
Check in OMNIC
-
Open OMNIC on the new PC.
-
Go to File → Open and browse to C:\My Documents\Omnic\Spectra.
-
Open a few files to confirm they work.
-
If the files open correctly, your data transfer is complete.