OMNIC Picta and Specta Printing issues in Windows 11
Issue
An error comes up when trying to print from Picta wizards (same sometimes also happen in Specta) in the priting dialog window.
Environment
OMNIC Picta, Specta
Resolution
1. Open Command Prompt with Admin rights
2. Copy and paste the command below and hit enter.
reg add "HKCU\Software\Microsoft\Print\UnifiedPrintDialog" /v "PreferLegacyPrintDialog" /d 1 /t REG_DWORD /f
If needed can be reversed by entering the command below.
reg delete "HKCU\Software\Microsoft\Print\UnifiedPrintDialog" /v "PreferLegacyPrintDialog" /f
Attachment(s)
| File | Last Modified |
|---|---|
| Picta issue when printing from Particle Wizard.png | March 28, 2025 |