OMNI Cell Liquid Volumes
This article provides the liquid volumes of the OMNI Cell liquid cell for the various spacers available.
Issue
What is the volume of the OMNI Cell liquid Cell for FTIR instruments
Environment
FT-IR Accessories
Resolution
The table below shows the volume in the liquid cell based on which spacer is being used.
| Spacer Thickness (mm) | Cell Volume (microliter) |
| 0.006 | 1.82 |
| 0.012 | 3.64 |
| 0.025 | 7.50 |
| 0.05 | 15.00 |
| 0.10 | 30.00 |
| 0.20 | 60.00 |
| 0.50 | 150.00 |
| 1.00 | 300.00 |