Omnic accessory experiment file keeps changing
Issue
Accessory experiment file keeps changing when smart accessory is inserted
Environment
- Nicolet iS20
- Nicolet iS10
- Nicolet Apex
- Nicolet iS50
Resolution
1. Check and clean the smart accessory chips contacts if they are dirty with a small a mount of Isopropyl alcohol
2. Check to see if postioning of Smart chip is correct.
See article Smart iTX Not Recognized by OMNIC - Thermo Fisher Scientific
Attachment(s)
| File | Last Modified |
|---|---|
| Smart Chip.png | February 11, 2025 |