Proper Stage Insert to use with Microcompression Cell II on the Nicolet RaptIR Microscope
Issue
When using the Microcompression Cell II (part 0045-434) with the RaptIR microscope, there is not enough distance when focused on the compression cell window to properly focus the condenser.
Environment
Nicolet RaptIR
Microcompression Cell II
Resolution
The correct stage insert must be used to allow for enough distance for proper focusing of both the z-stage and condenser.
Part 714-020400: iN10 Sample Holder - Compression Cell should be used.
Analysis
The Universal Sample Holder that ships with the Nicolet RaptIR microscope has a shallow recessed area for the Microcompression Cell II.
Because of the shallow recess, the z-stage is low when focusing on the sample, and this prevents the condenser from being able to achieve proper focus.
The iN10 Compression Cell Sample holder has a deeper recessed area for the compression cell, which allows the stage to be high enough for proper condenser focus to be achieved.
Attachment(s)
| File | Last Modified |
|---|---|
| RaptIR Universal Stage Insert.JPG | October 13, 2022 |
| Microcompression Cell II.JPG | October 13, 2022 |
| iN10 Compression Cell Stage Insert.JPG | October 13, 2022 |