Vials for use on the NIR Integrating Sphere
Issue
What vials are appropriate for use on the NIR integrating sphere?
Environment
Antaris NIR
Resolution
The diameter of the integrating sphere window is about 10 mm, so any flat bottom vials (also called "shell vials" or "tube vials") with diameters larger than 10 mm can be used.