iS5 or iS20 Failing valpro or Performance verification Wavelength or Error 1004 Laser Change fails
Issue
Nicolet iS5 Failing wavelength during Performance verification or Error 1004 Laser Change fails
Environment
- Nicolet iS5
- Nicolet iS20
Resolution
- To fix this issue run iS5 or iS20 Laser Verify H2O.mac by adding it to the Omnic tool bar from C:\my documents\omnic\Param\factory\LaserCal\
- After running Macro accept new laser Frequency and rerun Performance verification.