2I-01160 - Iss 2 - Advanced Solid Pack User Manual
Description:
Advanced solid pack user manual
The Advanced Solid Pack P/N GS01160 consists of the Specac Atlas™ Manual 15T Hydraulic Press P/N GS15011, offered together with a 13mm diameter Evacuable Pellet Die P/N GS03000 for the formation of 13mm diameter KBr pellets/discs typically used for the study of solid samples by transmission IR spectroscopy.
Environment:
- FT-IR
- Advanced Solid Pack
- Specac Atlas
Attachment(s):
| File | Last Modified |
|---|---|
| 2I-01160 - Iss 2 - Advanced Solid Pack User Manual.pdf | August 04, 2022 |