2I-31000 - Pearl Liquid Transmission Accessory - User Manual
Description:
The Pearl Liquid Transmission Accessory is designed for the qualitative and quantitative analysis of a very wide range of organic and aqueous based liquid sample types using the transmission technique of Infrared light passage through the sample. The liquid sample measurements are taken at ambient (room) temperatures and pressure conditions in a non-flowing environment. The principal features for the design allows for a quick, easy and consistent way of loading a liquid sample into a liquid cell assembly for a transmission spectroscopic measurement with an accurately known and repeatable pathlength for the liquid cell environment.
Environment:
Pearl Liquid Transmission Accessory
Attachment(s):
| File | Last Modified |
|---|---|
| 2I-31000 - Pearl Liquid Transmission Accessory - User Manual.pdf | August 04, 2022 |