350-013401 - VeeMax Variable Angle Specular Reflectance Accessory Installation and User's Guide
Description:
VeeMax variable angle specular reflectance accessory installation and user's guide
The PIKE Technologies VeeMax is a high throughput variable angle specular reflectanceaccessory designed for use in FTIR spectrometers. The design employs a unique optical design(patent applied for) which enables samples to be analyzed over a range of incident angles fromthirty to eighty degrees.
Environment:
- FT-IR
- VeeMax
Attachment(s):
| File | Last Modified |
|---|---|
| 350-013401 - VeeMax Variable Angle Specular Reflectance Accessory Installation and User's Guide.pdf | August 04, 2022 |