350-043001 - AutoDiffusIR Automated Diffuse Reflectance Accessory Installation and User Guide
Description:
Installation and User guide for the Automated Diffuse Reflectance Accessory
The PIKE Technologies AutoDiffusIR is high performance automated diffuse reflection accessory which is available for most FT-IR Spectrometers. The design employs a high efficiency fixed ellipsoidal reflector to collect the maximum amount of diffusely reflected energy from the sample. Operating under program control from within the FTIR software, the data generated may be processed either sequentially after each scan or stored to disk for further analysis.
Environment:
- FT-IR
- AutoDiffusIR
Attachment(s):
| File | Last Modified |
|---|---|
| 350-043001 - AutoDiffusIR Automated Diffuse Reflectance Accessory Installation and User Guide.pdf | August 04, 2022 |