350-0453000-01 - FlexIR Hollow Waveguide Accessory Installation and User Guide
Description:
FlexIR Hollow Waveguide Accessory installation and user guide
The FlexIR Hollow Waveguide Accessory is an excellent tool for remote and specific area analysis of a wide variety of samples. Visible surface contamination, small area material identification and bulky materials too large to fit into the FTIR sample compartment are a few of the many samples and application types for the FlexIR accessory.
Environment:
- FT-IR
- FlexIR
Attachment(s):
| File | Last Modified |
|---|---|
| 350-0453000-01 - FlexIR Hollow Waveguide Accessory Installation and User Guide.pdf | August 04, 2022 |