4A Molecular Sieve - Material Safety Data Sheet
Description:
This product is odorless, tan pellets, granules, beads, white powder or cake. This product is not combustible. When first wetted this product can heat up to the boiling point of water. Flooding with water will reduce the temperature.
Environment:
4A Molecular Sieve
Attachment(s):
| File | Last Modified |
|---|---|
| 4A Molecular Sieve - Material Safety Data Sheet.doc | August 04, 2022 |