700-0115 - Ver 2.9 - Econo-Press and Econo-Mount
Description:
This document contains information on the ECONO-PRESS.
The ECONO-PRESS is an economical die used to form KBr pellets for solid samples in infrared spectroscopy. The Model 0026-500 consists of three parts: one barrel (Pellet Form) and two polished forming bolts.
Environment:
- FT-IR
- Econo-Press
- Econo-Mount
Attachment(s):
| File | Last Modified |
|---|---|
| 700-0115 - Ver 2.9 - Econo-Press and Econo-Mount.pdf | August 04, 2022 |