CrushIR - Product Data Sheet
Description:
PIKE Technologies introduces a new and advanced hydraulic press for making excellent quality KBr pellets for transmission FTIR analysis. The CrushIR™ provides excellent reproducibility with its combination of digital pressure reading and adjustable maximum pressure setting thus preventing damage to the pellet die.
Environment:
CrushIR
FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| CrushIR - Product Data Sheet.pdf | August 04, 2022 |