DiffusIR - Product Data Sheet
Description:
The PIKE Technologies environmental chambers can be operated at temperatures ranging from -150 to 900 ºC and at pressures up to 1500 psi. The optional heat chambers are easily inserted and removed from the DiffusIR using push lock pins.
Environment:
DiffusIR
Attachment(s):
| File | Last Modified |
|---|---|
| DiffusIR - Product Data Sheet.pdf | August 04, 2022 |