FT-IR - OMNIC Tools
Description:
A Log File is used to store numerical output or processing information created through different OMNIC functions. It is an easy way to record numbers. In order to store information to a Log File, one must be open. A Log File can be opened under the File menu. If it is the first time the Log File command has been selected, OMNIC will ask for a new file name that the Log File will use to store the data. In the future this Log File can be selected or new ones can be created.