Falcon Mid-IR Transmission Accessory - Product Data Sheet
Description:
The PIKE Technologies mid-IR Falcon accessory is recommended for qualitative and quantitative analysis of liquids where it is necessary to control the temperature of the sample. Temperature range of the accessory is 5 °C to 130 °C with +/- 0.5% accuracy. Heating and cooling is controlled by a built-in Peltier device providing for reproducible ramping and for reaching target temperatures quickly and reliably. The system is driven by a Digital Temperature Controller – directly, or via PC.
Environment:
Falcon Mid-IR
Attachment(s):
| File | Last Modified |
|---|---|
| Falcon Mid-IR Transmission Accessory - Product Data Sheet.pdf | August 05, 2022 |