Heated NIR Transmission Accessory - Product Data Sheet
Description:
PIKE Technologies offers the Heated NIR Transmission Accessory for controlled temperature sampling. Applications include quantitative analysis of sample composition, especially where the sample needs to be at an elevated temperature to maintain its liquid state. Maintaining a precise temperature also improves the quantitative result by virtue of controlling the sample pathlength.
Environment:
Heated NIR Transmission Accessory
Attachment(s):
| File | Last Modified |
|---|---|
| Heated NIR Transmission Accessory - Product Data Sheet.pdf | August 04, 2022 |