MIRacle ATR - Product Data Sheet
Description:
The PIKE MIRacle™ is a universal ATR sampling accessory for analysis of solids, liquids, pastes, gels, and intractable materials. In its most popular configuration it is a single reflection ATR accessory with high IR throughput which makes it ideal for sample identification and QA/QC applications. Advanced options include three reflection ATR crystal plates to optimize for lower concentration components. Easily changeable crystal plate design provides analysis of a broad spectrum of sample types while ensuring constant sampling path length.
Environment:
MIRacle ATR
Attachment(s):
| File | Last Modified |
|---|---|
| MIRacle ATR - Product Data Sheet.pdf | August 04, 2022 |