Mid-IR IntegratIR Integrating Sphere Product Data Sheet
Description:
Mid-IR IntegratIR Integrating Sphere product information
The integrating sphere is very often an accessory of choice whenstudying reflectance properties of solids, analyzing light scatteringand/or highly absorbing samples and collecting spectra difficult toobtain with standard sampling techniques. PIKE Technologiesoffers Mid-IR integrating spheres, designed for research and standard applications that require high sensitivity and ability tocollect high quality data from difficult to analyze samples.
Environment:
- Mid-IR IntegratIR Integrating Sphere
- FT-IR
Attachment(s):
| File | Last Modified |
|---|---|
| Mid-IR IntegratIR Integrating Sphere Product Data Sheet.pdf | August 04, 2022 |