Polarizers - Product Data Sheet
Description:
PIKE Technologies offers polarizers for a wide variety of spectroscopy applications and are available in manual or automated versions. The polarizers are mounted in a standard 2" x 3" slide mount and are compatible with all FTIR spectrometers. Polarizer elements are 25 mm in diameter and, with mount, have a 20 mm clear aperture.
Environment:
Polarizers
Attachment(s):
| File | Last Modified |
|---|---|
| Polarizers - Product Data Sheet.pdf | August 04, 2022 |