Proteus Protein Analyzer - Product Specifications
Description:
The Proteus protein analyzer is a complete instrumentation and accessory package for the analysis of proteins. The package is based on Fourier transform infrared (FT-IR) technology and uses a Nicolet series FT-IR spectrometer to non-destructively scan protein samples for identification, structural analysis, defect analysis, impurity analysis and process quality monitoring.
Environment:
Proteus Protein
Attachment(s):
| File | Last Modified |
|---|---|
| Proteus Protein Analyzer - Product Specifications.pdf | August 04, 2022 |