Silver Gate Evolution - Data Sheet
Description:
The Silver Gate™ Evolution single reflection ATR system is a high throughput FTIR accessory capable of analysing solids, liquids and pastes with little or no sample preparation. The Silver Gate™ Evolution standard configuration has an interchangeable crystal plate with replacement crystal plates available. The sampling surface can be flat or recessed - depending on customer requirements and can be bought with or without the clamp bridge.
Environment:
Silver Gate Evolution
Attachment(s):
| File | Last Modified |
|---|---|
| Silver Gate Evolution - Data Sheet.pdf | August 04, 2022 |