AN 00029 - Two Case Studies of the Transfer of Near-Infrared Methods for the Analysis of Pharmaceutical Solid Dosage Forms
Description:
The ability to transfer calibration methods from a primary Fourier transform near-infrared (FT-NIR) instrument to a target instrument is assessed using two sets of tablets with varying physical characteristics.
Environment:
Antaris FT-NIR
Attachment(s):
| File | Last Modified |
|---|---|
| AN 00029 - Two Case Studies of the Transfer of Near-Infrared Methods for the Analysis of Pharmaceutical Solid Dosage Forms.pdf | July 25, 2022 |