AN 51262 - Automatic Power- Outage Recovery with an Antaris Process FT-NIR Analyzer and RESULT Software
Description:
One of the primary considerations in putting a Near-Infrared (NIR) analyzer online in a manufacturing process is how the analyzer will react in the event of a power outage on the factory grid. This document discusses the steps needed to set up an Antaris™FT-NIR process analyzer to recover automatically from a power outage. Once recovered, the instrument can be set to either begin collecting data automatically to monitor and record process information as the plant recovers, or it can be left in a “ready-to-run”state until other systems come online.
Environment:
- Antaris
- FT-NIR
Attachment(s):
| File | Last Modified |
|---|---|
| AN 51262 - Automatic Power- Outage Recovery with an Antaris Process FT-NIR Analyzer and RESULT Software.pdf | July 28, 2022 |