ISN 0001 - Advantages of FT-NIR
Description:
A near-infrared spectrum (12000-4000 cm-1) represents combination and overtone bands that are harmonics of absorption frequencies in the mid-infrared region. These combination and overtone bands correspond to the frequencies of vibrations between the bonds of the atoms making up the material. Because each different material is a unique combination of atoms, no two compounds produce the exact same near-infrared spectrum. Therefore, near-infrared spectroscopy can result in a positive identification (qualitative analysis) of each different material. In addition, the size of the peaks in the spectrum is a direct indication of the amount of material present.
Environment:
- FT-NIR
- NIR
Attachment(s):
| File | Last Modified |
|---|---|
| ISN 0001 - Advantages of FT-NIR.pdf | July 28, 2022 |