ISN 0007 Quantitative characterization of lactose crystalline forms
↵
Description:
Extended title: Quantitative Characterization of Lactose Crystalline Forms, A Case Study to Illustrate the Utility of FT-NIR as a Tool for the Analysis of the Crystalline Composition of Solid Materials
In this study, FT-NIR was used to quantitatively characterize binary and ternary mixtures of lactose, an important excipient used in many types of pharmaceutical dosage forms.
Environment:
- FT-NIR
- Antaris