ISN 0020 Method transfer through superior engineering
Description:
Analysis of variance related to user-replaceable components. The effect of replacing laser and source on the reproducibility of spectral data is discussed. The change in peak locations after replacing the laser and source are presented. The effect of replacing the source on line shape and band intensity is examined.