Modular Gas System
The Thermo Scientific™ Modular Gas System includes the FTIR Heated Valve Drawer for convenient gas selection and gas cell isolation. With the Heated Valve Drawer, you can monitor and control the temperature, pressure, and flow rate of your gas sample from sample point through the heated gas cell.
The Modular Gas System includes an open 19” rack mounted system and is designed to be used with the Thermo Scientific Antaris™ IGS analyzer, or the Nicolet™ iG™50, Nicolet iS™50, Nicolet iS10, or Nicolet iS20 spectrometers.
User guide
Find manuals for system components and options
269-145500 - Nicolet Antaris IGS Analyzer User Guide
269-308000 - Rev A - Gas Cell User Guide
269-153200 - Nicolet Antaris IGS Gas Cell User Guide