Focusing and Aligning the iN10 Microscope ATR Crystal
Description:
When using the ATR crystal on the iN10 microscope there are two main problems that can occur. One is low throughput and the second is the crystal not touching the proper spot on the sample. These problems can be fixed by focusing and alignment of the crystal. The attachment describes the process for adjusting the focus and alignment of the ATR crystal for the iN10.
Environment:
Nicolet iN10
Attachment(s):
| File | Last Modified |
|---|---|
| Focusing and Aligning the iN10 Microscope ATR Crystal.pdf | December 28, 2021 |