Optional Trinocular Eyepiece
Your microscope may have either a monocular eyepiece with only the camera or it may have the trinocular eyepiece with the camera and visual eyepieces.
| A | Visual eyepieces | Adjustable eyepieces for looking at the sample. Best used with the optional joystick. |
| B | Three-position view selector |
Controls the light path for the eyepieces.
|
| C | Camera | The USB camera is operated with OMNIC Paradigm software. |