Installing other accessories
Contains helpful supplementary information.
| Your accessory must be mounted on one of our baseplates. If it is not, you can remove an accessory from one of our baseplates and install your accessory on that baseplate. If you need assistance, contact us. |
- Remove any Smart Accessory, or if a Smart Accessory is not installed, remove the baseplate. Disconnect any cables that are connected at the back of the sample compartment.
- If the accessory is purged through the windows in the side of the sample compartment, install sidewall adapters.
- Install the accessory.
Use the instructions that came with the accessory to make any adjustments or perform any alignment procedures that are necessary.