Thermo Fisher Scientific
Installing sidewall adapters
Sidewall adapters are used to provide a tight purge seal for gas cells and other purged accessories. To install sidewall adapters, press the adapter ring toward the sidewall to compress the purge gasket, and then slide the adapter down into the clips on the side of the sample compartment.
114947_iS10 Install Sidewall Adapter.jpg
To remove a sidewall adapter, press the adapter ring toward the sidewall to compress the purge gasket, then slide the adapter up out of the clips on the side of the sample compartment.
  • Was this article helpful?