Maintaining detector dewar
If your instrument has a cooled detector, it will include a detector dewar. With proper care, a detector dewar should maintain its insulating vacuum for many years. If the vacuum leaks, the insulation will lose effectiveness and the following symptoms may occur:
- Liquid nitrogen boils off much faster than usual.
- The outside of the dewar stays cold more than 30 minutes after filling and may feel damp or become frosted.
- Water and atmospheric contaminants condensing on the detector window show up in spectra as unwanted peaks.
| If your instrument shows any of these symptoms, the detector dewar may have a vacuum leak. Contact us immediately for assistance. Leaving internal detector elements exposed to atmospheric pressure can permanently damage them. |