What options are available
Several accessories, or instruments, that expand what you can do with your spectrometer are available. The following is a list of some of these accessories. For information about additional accessories, contact our sales representative in your area.
| The accessory or instrument... | Does this... |
|
A Nicolet iZ10
|
Provides a second sampling area for routine analysis or for installation and usage of large, dedicated accessories like the TGA interface. |
|
An infrared microscope
|
Allows you to identify particles as small as 10 micrometers and perform many operations, such as surface analyses and chemical mapping. |
|
A near-IR integrating sphere
|
Uses a built-in detector to quickly identify materials contained in vials or blister packs. |
The following software is also available for your spectrometer:
- OMNIC Anywhere is cloud-based FT-IR software that allows you to analyze data away from the instrument, on any device, or share results with colleagues across the globe in a secure environment.
- TQ Analyst software provides an extensive suite of chemometrics features you can use to create methods to identify raw materials, perform quantitative analysis, and take spectral measurements.