Blue Touch-Point Setup
Description:
The blue “touch point” button located on the iS50 interface for iN5 is one in a series of touch points on the iS50 depending on the iS50 configuration. Touch points serve as a quick and easy way to select a specific beampath, load the related default experiment file and start a data collection. The attached procedure describes how to set up the touch point button.
Environment:
- Nicolet iN5
- Nicolet iS50
Attachment(s):
| File | Last Modified |
|---|---|
| How to Setup the iS50 Interface “Blue Touch-Point “to Switch to the iN5 Microscope Beam Path.pdf | December 18, 2021 |