Mosaic Image Acquisitions
Description:
Mosaic Images will not align/display correctly.
Environment:
- Noran System Six
- NSS
- NS6
Resolution:
When Mosaic images are performed, we need to assure that when the stage is moved in X direction, that the image on the NSS does NOT move at all in the Y direction.