NSS software opens with errors and also behaves irregularly.
Description:
NSS Software Opens With Errors About Inability To Communicate With Other Microanalysis Devices
Environment:
- Noran System 6
- Noran Ssytem 7
- Pathfinder
RESOLUTION:
- ASSURE any optional equipment running with the NSS is turned on
- Ensure a valid ethernet connection is in place. This optional equipment includes WDS, EBSD, and Detector Protection System (Low Mag &Vent Lock).
- Attached is examples of NSS system component IP Addresses.
Attachment(s):
File | Last Modified |
---|---|
NSS System IP Addresses.jpg | June 03, 2022 |