Thermo Fisher Scientific
Creating a mosaic image
Description: 
I cannot create an image montage
Environment:
  • Noran System 6
  • Noran System 7
Resolution:
  1. Multiple images can be stitched together in NSS software. However, only images collected using Analysis Automation can be stitched together.
  2. Manually collected images cannot be stitched together.
  • Was this article helpful?