Thermo Fisher Scientific
Mosaic Image Acquisitions
Description: 
Mosaic Images will not align/display correctly.
Environment:
  • Noran System Six
  • NSS
  • NS6
Resolution:
When Mosaic images are performed, we need to assure that when the stage is moved in X direction, that the image on the NSS does NOT move at all in the Y direction.
  • Was this article helpful?