Thermo Fisher Scientific
NSS software opens with errors and also behaves irregularly.
Description: 
NSS Software Opens With Errors About Inability To Communicate With Other Microanalysis Devices
Environment:
  • Noran System 6
  • Noran Ssytem 7
  • Pathfinder
RESOLUTION:
  1. ASSURE any optional equipment running with the NSS is turned on
  2. Ensure a valid ethernet connection is in place. This optional equipment includes WDS, EBSD, and Detector Protection System (Low Mag &Vent Lock).
  3. Attached is examples of NSS system component IP Addresses.
  • Was this article helpful?