Thermo Fisher Scientific
Obtaining DPPCal2 troubleshooting data from NS7 EDS install/calibration.
Description: 
Irregular results are seen when calibrating an EDS detecter on an NS7 system
Environment:
  • Noran Ssytem 7
Resolution:
  1. NS7 has the ability to store-save-amend crucial DPP parameters in csv files for review by engineers at the factory to aid in resolving irregular calibration issues.  
  2. Location of this data is in the Shared Docs folder in a  separate "DPPCal Data" folder.
  • Was this article helpful?