Liquid nitrogen
Because the microscope uses a cooled detector, you need a supply of liquid nitrogen to cool the detector element.
|
Avoid hazard The gas boiling off liquid nitrogen can create an oxygen deficient environment in an insufficiently ventilated room. |
Avoid hazard Be careful not to contact the liquid nitrogen with your skin. It is extremely cold. Exposure may cause burns. Wear protective gloves and goggles and follow |