Thermo Fisher Scientific
Weight and dimensions
Weight
Your microscope weighs a maximum of 70.3 kg (155 lb).
Make sure your work surface can support this much weight in addition to the weight of the Nicolet iS50 spectrometer, the system computer, and any accessories you are using.
Dimensions
The dimensions of your microscope are shown below.
  • Wide: 45 cm (17.7 in)
  • Tall: 49.5 cm (19.5 in)
  • Deep: 71.7 cm (28.2 in)
Dimensions of the Nicolet RaptIR FTIR Microscope
Dimensions of the iS50 spectrometer 
iS50 spectrometer base dimensions
• Weight: 64 kg (140 lbs)
• Width: 610 mm (24 in)
• Depth: 711 mm (28 in)
• Height: 280 mm (11 in)
iS50 spectrometer with the ABX Dimensions
• Weight: 68 kg (149 lbs)
• Width: 610 mm (24 in)
• Depth: 711 mm (28 in)
• Height: 508 mm (20 in)
Combined maximum dimensions 
• Weight: 138.3 kg (305 lbs)
• Width: 114 cm (45 in)
• Depth: 71.7 cm (28 in)
• Height: 51 cm (20 in)
Clearances
Leave space around and behind the system to reach the cable and purge connections. Leaving additional space around the instrument so that it is easier to access will also make service calls faster and easier.
  • Was this article helpful?