Thermo Fisher Scientific
Data Security and Integrity with OMNIC Paradigm Software
To ensure the security and integrity of your data and to help your lab comply with 21 CFR Part 11 or other regulations, pair the OMNIC™ Paradigm software with Thermo Scientific Security Suite software. Security Suite is a comprehensive data security toolset, allowing you to perform the following tasks:
  • Limit and control access to features and tools in the OMNIC Paradigm software and other instrument applications.
  • Set and manage security policies to ensure proper data handling.
  • Customize and enforce digital signatures to establish responsibility.
  • Log and view security events. You can purchase Security Suite software to manage a single system or to manage multiple instruments distributed on a network. After installation, configuring the OMNIC Paradigm application for security takes only moments. For a discussion of how Security Suite software can help you comply with 21 CFR Part 11, see 21 CFR Part 11 Compliance.
  • Was this article helpful?