Maximum Number of Characters for a Measurement Tag in OMNIC Paradigm Software
This article explains that the maximum number of characters allowed for a measurement Tag in OMNIC Paradigm is 64 characters. Exceeding this length for the tag can cause various problems and a Navigation Failed error message.
Issue
There is a maximum number of characters (64) allowed for a measurement Tag in OMNIC Paradigm.
If the length is exceeded, a few different problems might occur.
- A red box will appear around the Tag field on the preview screen or the Dashboard
- The collection buttons on the Dashboard will be disabled
- The Start Sample Measurement button on the preview screen will be disabled
- An error may display when trying to Preview a measurement that says "Navigation Failed. Check measurement settings and try again."
Environment
OMNIC Paradigm
Resolution
Always use 64 characters or less for any measurement Tag in the OMNIC Paradigm software.
Attachment(s)
| File | Last Modified |
|---|---|
| Preview Window Tag too Long.jpg | December 23, 2025 |
| Dashboard - Tag too long.jpg | December 23, 2025 |